siRNA Details
| Gene Symbol: Shbg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012650.1 | NP_036782.1 | 1, 2 | 140 | ||
| XM_006246586.2 | XP_006246648.1 | 2, 3 | 889 | |||
| GenBank mRNA | BC097336.1 | - | 1, 2 | 128 | 7453185 | |
| M15034.1 | - | 1, 2 | 140 | |||
| M31179.1 | - | 1, 2 | 92 | |||

Shbg
Genomic Map