siRNA Details
| Gene Symbol: Tgfa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012671.2 | NP_036803.1 | 6 | 1176 | ||
| XM_008763050.1 | XP_008761272.1 | 6 | 2070 | |||
| GenBank mRNA | M31076.1 | - | 6 | 1177 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tgfa| Gene Symbol: Tgfa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012671.2 | NP_036803.1 | 6 | 1176 | ||
| XM_008763050.1 | XP_008761272.1 | 6 | 2070 | |||
| GenBank mRNA | M31076.1 | - | 6 | 1177 | ||