siRNA Details
| Gene Symbol: Vcsa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012684.4 | NP_036816.4 | 3 | 226 | ||
| GenBank mRNA | A07543.1 | - | 3 | 233 | ||
| CB769246.1 | - | 3 | 238 | |||
| M63112.1 | - | 3 | 233 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vcsa1| Gene Symbol: Vcsa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012684.4 | NP_036816.4 | 3 | 226 | ||
| GenBank mRNA | A07543.1 | - | 3 | 233 | ||
| CB769246.1 | - | 3 | 238 | |||
| M63112.1 | - | 3 | 233 | |||