siRNA Details
| Gene Symbol: Tacr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080768.1 | NP_542946.1 | 4 | 1285 | ||
| GenBank mRNA | M31838.1 | - | 4 | 1285 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tacr2| Gene Symbol: Tacr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080768.1 | NP_542946.1 | 4 | 1285 | ||
| GenBank mRNA | M31838.1 | - | 4 | 1285 | ||
Set Membership: |
|