siRNA Details
| Gene Symbol: Stx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012748.1 | NP_036880.1 | 2 | 43 | ||
| XM_006249249.2 | XP_006249311.1 | 2 | 201 | |||
| XM_006249250.2 | XP_006249312.1 | 2 | 201 | |||
| XM_006249253.2 | XP_006249315.1 | 3 | 1947 | |||
| XM_008769188.1 | XP_008767410.1 | 3 | 2414 | |||
| GenBank mRNA | AY302700.1 | - | 2 | 43 | ||
| L20823.1 | - | 2 | 43 | |||
| L20888.1 | - | 2 | 43 | |||
| L20889.1 | - | 2 | 43 | |||

Stx2
Genomic Map