siRNA Details
| Gene Symbol: Apod | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012777.1 | NP_036909.1 | 3 | 207 | ||
| GenBank mRNA | FQ212999.1 | - | 3 | 207 | ||
| X55572.1 | - | 3 | 207 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Apod| Gene Symbol: Apod | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012777.1 | NP_036909.1 | 3 | 207 | ||
| GenBank mRNA | FQ212999.1 | - | 3 | 207 | ||
| X55572.1 | - | 3 | 207 | |||