siRNA Details
| Gene Symbol: Pdgfa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012801.1 | NP_036933.1 | 7 | 1036 | ||
| XM_006248918.2 | XP_006248980.1 | 5 | 1018 | |||
| XM_006248919.2 | XP_006248981.1 | 5 | 1018 | |||
| XM_008768940.1 | XP_008767162.1 | 6 | 2617 | |||
| GenBank mRNA | BC061731.1 | - | 5 | 1023 | 5598667 | |
| D10106.1 | - | 5 | 595 | |||
| L06238.1 | - | 7 | 1036 | |||
| L06894.1 | - | 6 | 680 | |||
| S57864.1 | - | 3 | 381 | |||
| Z14120.1 | - | 5 | 530 | |||

Pdgfa
Genomic Map