siRNA Details
| Gene Symbol: Egf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012842.1 | NP_036974.1 | 1422 | |||
| GenBank mRNA | BC062030.1 | - | 1407 | 5623849 | ||
| JX149564.1 | - | 1266 | ||||
| JX437943.1 | - | 1266 | ||||
| U04842.1 | - | 1422 | ||||
| Gene Symbol: LOC100910178 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008760814.1 | XP_008759036.1 | 6 | 1433 | ||
| XM_008760815.1 | XP_008759037.1 | 6 | 1432 | |||
| XM_008760816.1 | XP_008759038.1 | 6 | 1193 | |||

Egf
Genomic Map