siRNA Details
| Gene Symbol: Egf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012842.1 | NP_036974.1 | 1996 | |||
| GenBank mRNA | BC062030.1 | - | 1978 | 5623849 | ||
| JX149564.1 | - | 1840 | ||||
| JX437943.1 | - | 1840 | ||||
| U04842.1 | - | 1996 | ||||
| Gene Symbol: LOC100910178 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008760814.1 | XP_008759036.1 | 11 | 2004 | ||
| XM_008760815.1 | XP_008759037.1 | 11 | 2003 | |||
| XM_008760816.1 | XP_008759038.1 | 11 | 1764 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_010113, BC060741, AK085707, AK157778, V00741, and others.

Egf
Genomic Map