siRNA Details
| Gene Symbol: Emp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012843.2 | NP_036975.1 | 386 | |||
| XM_006237515.2 | XP_006237577.1 | 495 | ||||
| XM_008763369.1 | XP_008761591.1 | 495 | ||||
| GenBank mRNA | BC087031.1 | - | 386 | 7192247 | ||
| Z54212.1 | - | 355 | ||||

Emp1
Genomic Map