siRNA Details
| Gene Symbol: Hsd17b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012851.2 | NP_036983.1 | 2 | 290 | ||
| GenBank mRNA | BC086365.1 | - | 2 | 275 | 7305002 | |
| X78811.1 | - | 2 | 251 | |||
| X97754.1 | - | 2 | 291 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hsd17b1| Gene Symbol: Hsd17b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012851.2 | NP_036983.1 | 2 | 290 | ||
| GenBank mRNA | BC086365.1 | - | 2 | 275 | 7305002 | |
| X78811.1 | - | 2 | 251 | |||
| X97754.1 | - | 2 | 291 | |||