siRNA Details
| Gene Symbol: Timp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012886.2 | NP_037018.2 | 3, 4 | 613 | ||
| XM_006241162.2 | XP_006241224.1 | 3, 4 | 642 | |||
| GenBank mRNA | BC097335.1 | - | 3, 4 | 613 | 7443686 | |
| U27201.1 | - | 3, 4 | 328 | |||

Timp3
Genomic Map