siRNA Details
| Gene Symbol: Atpif1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012915.2 | NP_037047.2 | 3 | 272 | ||
| GenBank mRNA | BC126065.1 | - | 3 | 237 | 7442425 | |
| D13122.1 | - | 3 | 272 | |||
| FQ221416.1 | - | 3 | 257 | |||
| FQ229412.1 | - | 3 | 234 | |||

Atpif1
Genomic Map