siRNA Details
| Gene Symbol: Cpt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012930.1 | NP_037062.1 | 4 | 441 | ||
| GenBank mRNA | J05470.1 | - | 4 | 441 | ||
| U88295.1 | - | 4 | 379 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cpt2| Gene Symbol: Cpt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012930.1 | NP_037062.1 | 4 | 441 | ||
| GenBank mRNA | J05470.1 | - | 4 | 441 | ||
| U88295.1 | - | 4 | 379 | |||