siRNA Details
| Gene Symbol: Kcne1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012973.1 | NP_037105.1 | 2 | 388 | ||
| XM_006248034.2 | XP_006248096.1 | 2 | 401 | |||
| XM_006248035.2 | XP_006248097.1 | 2 | 555 | |||
| XM_008768566.1 | XP_008766788.1 | 2 | 772 | |||
| GenBank mRNA | M22412.1 | - | 2 | 388 | ||
| M36461.1 | - | 1 | 365 | |||

Kcne1
Genomic Map