siRNA Details
Gene Symbol: Myo1e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173101.1 | NP_775124.1 | 13 | 1788 | ||
GenBank mRNA | X74815.1 | - | 13 | 1788 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Myo1e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173101.1 | NP_775124.1 | 13 | 1788 | ||
GenBank mRNA | X74815.1 | - | 13 | 1788 |