siRNA Details
Gene Symbol: Ccl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013025.2 | NP_037157.2 | 3 | 712 | ||
GenBank mRNA | U22414.1 | - | 3 | 712 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ccl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013025.2 | NP_037157.2 | 3 | 712 | ||
GenBank mRNA | U22414.1 | - | 3 | 712 |