siRNA Details
| Gene Symbol: Ube2i | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013050.1 | NP_037182.1 | 5 | 336 | ||
| XM_006245918.2 | XP_006245980.1 | 5 | 336 | |||
| XM_006245919.2 | XP_006245981.1 | 5 | 1016 | |||
| XM_006245920.2 | XP_006245982.1 | 5 | 411 | |||
| XM_008767549.1 | XP_008765771.1 | 5 | 468 | |||
| GenBank mRNA | BC086324.1 | - | 5 | 334 | 7305067 | |
| BC086592.1 | - | 5 | 334 | 7310403 | ||
| FQ226760.1 | - | 5 | 411 | |||
| FQ228426.1 | - | 5 | 321 | |||
| FQ229169.1 | - | 5 | 321 | |||
| U54632.1 | - | 5 | 336 | |||

Ube2i
Genomic Map