siRNA Details
| Gene Symbol: Ptger1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278475.1 | NP_001265404.1 | 2 | 1110 | ||
| XM_008772362.1 | XP_008770584.1 | 2 | 1762 | |||
| GenBank mRNA | D88751.1 | - | 1 | 1402 | ||
| D88752.1 | - | 1 | 1402 | |||
| U68037.1 | - | 1 | 818 | |||
More Information
Set Membership: |
|

Ptger1
Genomic Map