siRNA Details
| Gene Symbol: Cd28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013121.1 | NP_037253.1 | 1 | 78 | ||
| GenBank mRNA | X55288.1 | - | 1 | 78 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cd28| Gene Symbol: Cd28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013121.1 | NP_037253.1 | 1 | 78 | ||
| GenBank mRNA | X55288.1 | - | 1 | 78 | ||