siRNA Details
| Gene Symbol: Tgfb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013174.2 | NP_037306.1 | 4 | 1113 | ||
| XM_006240366.1 | XP_006240428.1 | 5 | 1457 | |||
| GenBank mRNA | BC092195.1 | - | 4 | 1100 | 7313608 | |
| U03491.1 | - | 4 | 1113 | |||

Tgfb3
Genomic Map