siRNA Details
| Gene Symbol: Aqp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022181.1 | NP_071517.1 | 4 | 907 | ||
| GenBank mRNA | AF083879.1 | - | 4 | 907 | ||
| L28113.1 | - | 4 | 935 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Aqp6| Gene Symbol: Aqp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022181.1 | NP_071517.1 | 4 | 907 | ||
| GenBank mRNA | AF083879.1 | - | 4 | 907 | ||
| L28113.1 | - | 4 | 935 | |||