siRNA Details
| Gene Symbol: Ptma | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021740.1 | NP_068508.1 | 5 | 626 | ||
| GenBank mRNA | M86564.1 | - | 5 | 626 | ||
| X55326.1 | - | 5 | 626 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ptma| Gene Symbol: Ptma | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021740.1 | NP_068508.1 | 5 | 626 | ||
| GenBank mRNA | M86564.1 | - | 5 | 626 | ||
| X55326.1 | - | 5 | 626 | |||