siRNA Details
| Gene Symbol: Nfib | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031566.1 | NP_113754.1 | 11 | 1536 | ||
| XM_006238335.2 | XP_006238397.1 | 12 | 2317 | |||
| XM_006238336.2 | XP_006238398.1 | 12 | 2290 | |||
| XM_006238339.2 | XP_006238401.1 | 9 | 1977 | |||
| XM_008763762.1 | XP_008761984.1 | 12 | 2032 | |||
| GenBank mRNA | AB012230.2 | - | 12 | 1625 | ||
| AB012231.2 | - | 11 | 1536 | |||
| AB012232.2 | - | 9 | 1314 | |||

Nfib
Genomic Map