siRNA Details
| Gene Symbol: Spt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019171.1 | NP_062044.1 | 4 | 375 | ||
| GenBank mRNA | M33976.1 | - | 4 | 375 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Spt1| Gene Symbol: Spt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019171.1 | NP_062044.1 | 4 | 375 | ||
| GenBank mRNA | M33976.1 | - | 4 | 375 | ||