siRNA Details
| Gene Symbol: Cyp2c7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017158.2 | NP_058854.1 | 9 | 1416 | ||
| GenBank mRNA | BC097939.1 | - | 9 | 1399 | 7368637 | |
| BC127503.1 | - | 9 | 1455 | 7371672 | ||
| M18335.1 | - | 9 | 1417 | |||
| M31031.1 | - | 9 | 1374 | |||
| Gene Symbol: LOC100361547 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008759635.1 | XP_008757857.1 | 9 | 1272 | ||

Cyp2c7
Genomic Map