siRNA Details
| Gene Symbol: Cyp2c7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017158.2 | NP_058854.1 | 9 | 1365 | ||
| XM_008759609.1 | XP_008757831.1 | 9 | 1487 | |||
| GenBank mRNA | BC097939.1 | - | 9 | 1348 | 7368637 | |
| BC127503.1 | - | 9 | 1404 | 7371672 | ||
| FQ218330.1 | - | 8 | 2401 | |||
| M18335.1 | - | 9 | 1366 | |||
| M31031.1 | - | 9 | 1323 | |||
| Gene Symbol: LOC100361547 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008759635.1 | XP_008757857.1 | 9 | 1221 | ||

Cyp2c7
Genomic Map