siRNA Details
| Gene Symbol: C5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001317163.1 | NP_001304092.1 | 33 | 4552 | ||
| NM_001735.2 | NP_001726.2 | 33 | 4226 | |||
| XM_011518980.2 | XP_011517282.1 | 34 | 4595 | |||
| XM_017015102.1 | XP_016870591.1 | 35 | 4747 | |||
| XM_017015103.1 | XP_016870592.1 | 33 | 4380 | |||
| GenBank mRNA | AB209031.1 | - | 33 | 4550 | ||
| BC113738.1 | - | 33 | 4221 | 8322790 | ||
| BC113740.1 | - | 33 | 4221 | 8322792 | ||
| M57729.1 | - | 33 | 4208 | |||
| M65134.1 | - | 23 | 2963 | |||

C5
Genomic Map