siRNA Details
| Gene Symbol: TPPP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007030.2 | NP_008961.1 | 3 | 456 | ||
| XM_005248237.3 | XP_005248294.2 | 3 | 521 | |||
| XM_017008993.1 | XP_016864482.1 | 3 | 2652 | |||
| XM_017008994.1 | XP_016864483.1 | 5 | 507 | |||
| GenBank mRNA | AB017016.1 | - | 3 | 367 | ||
| AK054920.1 | - | 2 | 1053 | |||
| AK098392.1 | - | 3 | 412 | |||
| AY319261.1 | - | 2 | 334 | |||
| BC131506.1 | - | 2 | 352 | 40082752 | ||
| BC152382.1 | - | 2 | 337 | 100013441 | ||
| BT020035.1 | - | 2 | 334 | |||
| DB478250.2 | - | 3 | 456 | |||

TPPP
Genomic Map