siRNA Details
| Gene Symbol: KERA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007035.3 | NP_008966.1 | 2 | 1399 | ||
| GenBank mRNA | AF063301.1 | - | 1 | 788 | ||
| AF205403.1 | - | 2 | 1028 | |||
| AK314990.1 | - | 2 | 961 | |||
| BC032667.2 | - | 1 | 789 | 5541591 | ||

KERA
Genomic Map