siRNA Details
| Gene Symbol: ESM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007036.4 | NP_008967.1 | 2 | 483 | ||
| GenBank mRNA | AK311817.1 | - | 2 | 490 | ||
| BC011989.1 | - | 2 | 473 | 3882426 | ||
| BM423977.1 | - | 2 | 560 | |||
| X89426.1 | - | 2 | 469 | |||

ESM1
Genomic Map