siRNA Details
| Gene Symbol: ESM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135604.1 | NP_001129076.1 | 1 | 173 | ||
| NM_007036.4 | NP_008967.1 | 1 | 173 | |||
| GenBank mRNA | AJ973643.1 | - | 1 | 120 | ||
| AK300634.1 | - | 1 | 180 | |||
| AK311817.1 | - | 1 | 180 | |||
| BC011989.1 | - | 1 | 163 | 3882426 | ||
| BM423977.1 | - | 1 | 250 | |||
| X89426.1 | - | 1 | 159 | |||

ESM1
Genomic Map