siRNA Details
| Gene Symbol: C6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000065.3 | NP_000056.2 | 17 | 2667 | ||
| NM_001115131.2 | NP_001108603.2 | 17 | 2694 | |||
| XM_005248357.2 | XP_005248414.1 | 17 | 2742 | |||
| XM_006714496.3 | XP_006714559.1 | 18 | 2799 | |||
| XM_011514114.2 | XP_011512416.1 | 18 | 2826 | |||
| XM_011514115.2 | XP_011512417.1 | 18 | 2747 | |||
| XM_011514116.2 | XP_011512418.1 | 19 | 2863 | |||
| XM_011514117.2 | XP_011512419.1 | 19 | 2905 | |||
| XM_011514118.2 | XP_011512420.1 | 18 | 2643 | |||
| XM_011514119.2 | XP_011512421.1 | 17 | 2769 | |||
| XM_011514121.2 | XP_011512423.1 | 13 | 1912 | |||
| XM_017009818.1 | XP_016865307.1 | 19 | 2886 | |||
| XM_017009819.1 | XP_016865308.1 | 14 | 2089 | |||
| GenBank mRNA | AB126592.1 | - | 16 | 2429 | ||
| AK292509.1 | - | 17 | 2565 | |||
| AK313673.1 | - | 17 | 2586 | |||
| BC035723.1 | - | 17 | 2694 | 5591446 | ||
| J05024.1 | - | 17 | 2486 | |||
| J05064.1 | - | 17 | 2584 | |||

C6
Genomic Map