siRNA Details
| Gene Symbol: PRDM7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001098173.1 | NP_001091643.1 | 10 | 2096 | ||
| XM_005256274.3 | XP_005256331.1 | 11 | 2275 | |||
| XM_011522829.1 | XP_011521131.1 | 10 | 1992 | |||
| GenBank mRNA | AF274347.1 | - | 5 | 1511 | ||
| AF274348.1 | - | 3 | 1316 | |||
| AL137711.1 | - | 3 | 1141 | |||

PRDM7
Genomic Map