siRNA Details
| Gene Symbol: EMILIN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007046.3 | NP_008977.1 | 4 | 2176 | ||
| GenBank mRNA | AF088916.1 | - | 4 | 1677 | ||
| AL050138.1 | - | 4 | 1607 | |||
| BC009947.2 | - | 1 | 371 | |||
| BC136279.1 | - | 4 | 1776 | 9020266 | ||
| BC142638.1 | - | 4 | 904 | |||
| BC142688.1 | - | 4 | 2058 | 40148654 | ||

EMILIN1
Genomic Map