siRNA Details
| Gene Symbol: BTN3A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145008.1 | NP_001138480.1 | 10 | 1534 | ||
| NM_001145009.1 | NP_001138481.1 | 10 | 2447 | |||
| NM_007048.5 | NP_008979.3 | 10 | 1690 | |||
| NM_194441.2 | NP_919423.1 | 10 | 1984 | |||
| XM_005248833.2 | XP_005248890.1 | 9 | 1637 | |||
| XM_005248834.3 | XP_005248891.1 | 10 | 2149 | |||
| XM_006714983.2 | XP_006715046.1 | 9 | 1395 | |||
| GenBank mRNA | AK223474.1 | - | 9 | 1471 | ||
| AK290193.1 | - | 10 | 1687 | |||
| AK298652.1 | - | 10 | 1419 | |||
| AK299327.1 | - | 10 | 1476 | |||
| BC118586.1 | - | 10 | 1651 | 40085016 | ||
| BC121800.1 | - | 10 | 1651 | 40085015 | ||
| CB148209.1 | - | 6 | 425 | |||
| DA044176.1 | - | 1 | 438 | |||
| U90552.2 | - | 10 | 1620 | |||

BTN3A1
Genomic Map