siRNA Details
| Gene Symbol: Obox3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145707.3 | NP_663753.3 | 5 | 1518 | ||
| GenBank mRNA | AK139750.1 | - | 5 | 1519 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Obox3| Gene Symbol: Obox3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145707.3 | NP_663753.3 | 5 | 1518 | ||
| GenBank mRNA | AK139750.1 | - | 5 | 1519 | ||