siRNA Details
| Gene Symbol: Kif28p | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270385.1 | NP_001257314.1 | 23 | 3058 | ||
| GenBank mRNA | AB573712.1 | - | 23 | 3058 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kif28p| Gene Symbol: Kif28p | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270385.1 | NP_001257314.1 | 23 | 3058 | ||
| GenBank mRNA | AB573712.1 | - | 23 | 3058 | ||