siRNA Details
| Gene Symbol: Olfr723 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011530.2 | NP_001011530.2 | 1 | 156 | ||
| GenBank mRNA | BC150980.1 | - | 1 | 156 | 9087893 | |
| Gene Symbol: Olfr724 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146492.2 | NP_666703.2 | 1 | 156 | ||
| GenBank mRNA | BC120628.1 | - | 1 | 156 | 40129551 | |
| BC125538.1 | - | 1 | 156 | 40130053 | ||

Olfr723
Genomic Map