siRNA Details
| Gene Symbol: Vmn1r160 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166724.1 | NP_001160196.1 | 1 | 431 | ||
| Gene Symbol: Vmn1r123 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166707.1 | NP_001160179.1 | 1 | 431 | ||

Vmn1r160
Genomic Map