siRNA Details
| Gene Symbol: Smok3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001126045.1 | NP_001119517.1 | 2 | 660 | ||
| NM_001168602.1 | NP_001162073.1 | 2 | 813 | |||
| GenBank mRNA | AJ245454.1 | - | 2 | 813 | ||
| BC150960.1 | - | 660 | 9087871 | |||
| Gene Symbol: Smok3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039889.3 | NP_001034978.1 | 2 | 834 | ||
| GenBank mRNA | AK133180.1 | - | 2 | 835 | ||

Smok3a
Genomic Map