siRNA Details
| Gene Symbol: Atp5j2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC029226.1 | - | 1, 2 | 132 | 2651401 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Atp5j2| Gene Symbol: Atp5j2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC029226.1 | - | 1, 2 | 132 | 2651401 | |