siRNA Details
| Gene Symbol: BVES | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199563.1 | NP_001186492.1 | 3, 4 | 575 | ||
| NM_007073.4 | NP_009004.2 | 3, 4 | 445 | |||
| NM_147147.3 | NP_671488.1 | 3, 4 | 515 | |||
| XM_011535398.2 | XP_011533700.1 | 3, 4 | 450 | |||
| GenBank mRNA | AF204172.1 | - | 3, 4 | 469 | ||
| AK289979.1 | - | 3, 4 | 559 | |||
| BC034425.1 | - | 3, 4 | 515 | 4821891 | ||
| BC040502.1 | - | 3, 4 | 575 | 5295907 | ||
 Back To Top
 Back To Top
 BVES
BVES Genomic Map
Genomic Map Back To Top
 Back To Top