siRNA Details
| Gene Symbol: Gm8369 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164202.1 | NP_001157674.1 | 3 | 260 | ||
| GenBank mRNA | AK156452.1 | - | 4 | 385 | ||
| BB637086.1 | - | 3 | 260 | |||
| BE861164.1 | - | 2 | 99 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm8369| Gene Symbol: Gm8369 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164202.1 | NP_001157674.1 | 3 | 260 | ||
| GenBank mRNA | AK156452.1 | - | 4 | 385 | ||
| BB637086.1 | - | 3 | 260 | |||
| BE861164.1 | - | 2 | 99 | |||