siRNA Details
| Gene Symbol: Pgam1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023418.2 | NP_075907.2 | 4 | 1804 | ||
| GenBank mRNA | AF283667.1 | - | 4 | 1710 | ||
| AK004921.1 | - | 4 | 1660 | |||
| AK009905.2 | - | 4 | 1657 | |||
| AK144681.1 | - | 4 | 1658 | |||
| AK150716.1 | - | 4 | 1653 | |||
| AK152443.1 | - | 4 | 1653 | |||
| AK153202.1 | - | 4 | 1657 | |||
| AK159305.1 | - | 4 | 1657 | |||
| AK165410.1 | - | 4 | 1653 | |||
| AK168949.1 | - | 4 | 1650 | |||
| BC002241.1 | - | 4 | 1746 | 3491964 | ||
| BC005661.1 | - | 4 | 1712 | 3588761 | ||
| BC066844.1 | - | 4 | 1641 | 30471755 | ||
| BC067412.1 | - | 3 | 1457 | |||
| BC083090.1 | - | 4 | 1652 | 30444229 | ||
| BC106139.1 | - | 4 | 1724 | 6528521 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: BC065582.

Pgam1
Genomic Map