siRNA Details
| Gene Symbol: CORO1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001193333.2 | NP_001180262.1 | 6, 7 | 1066 | ||
| NM_007074.3 | NP_009005.1 | 5, 6 | 948 | |||
| XM_011545714.2 | XP_011544016.1 | 5, 6 | 761 | |||
| XM_017022885.1 | XP_016878374.1 | 5, 6 | 936 | |||
| XM_017022886.1 | XP_016878375.1 | 5, 6 | 758 | |||
| GenBank mRNA | AB209221.1 | - | 2, 3 | 2893 | ||
| AF495470.1 | - | 5, 6 | 753 | |||
| AK096332.1 | - | 4, 5 | 1943 | |||
| AK097964.1 | - | 1, 2 | 537 | |||
| AK123401.1 | - | 2, 3 | 1192 | |||
| AK314714.1 | - | 6, 7 | 867 | |||
| BC110374.1 | - | 5, 6 | 765 | 5733853 | ||
| BC126385.1 | - | 5, 6 | 652 | 8992101 | ||
| BC126387.1 | - | 5, 6 | 652 | 8992103 | ||
| D44497.1 | - | 5, 6 | 731 | |||
| U34690.1 | - | 5, 6 | 723 | |||
| X89109.1 | - | 5, 6 | 724 | |||

CORO1A
Genomic Map