siRNA Details
| Gene Symbol: Igh-V3609N | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | D29669.1 | - | 1 | 34 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Igh-V3609N| Gene Symbol: Igh-V3609N | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | D29669.1 | - | 1 | 34 | ||