siRNA Details
| Gene Symbol: RGD1564897 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_578467.2 | XP_578467.1 | 1 | 186 | ||
| Gene Symbol: LOC100912356 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749975.2 | XP_003750023.1 | 1 | 186 | ||

RGD1564897
Genomic Map