siRNA Details
| Gene Symbol: RGD1565363 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_002728805.3 | XP_002728851.1 | 1 | 255 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1565363| Gene Symbol: RGD1565363 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_002728805.3 | XP_002728851.1 | 1 | 255 | ||