siRNA Details
| Gene Symbol: Tmem52b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109560.1 | NP_001103030.1 | 4 | 735 | ||
| XM_006237102.2 | XP_006237164.1 | 5 | 672 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem52b| Gene Symbol: Tmem52b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109560.1 | NP_001103030.1 | 4 | 735 | ||
| XM_006237102.2 | XP_006237164.1 | 5 | 672 | |||